Assay Details
Gene Symbol: Serpinb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057108.2 | NP_476449.1 | 4 - 5 | 536 | 112 | |
XM_008769405.1 | XP_008767627.1 | 3 - 4 | 694 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
