Assay Details
Gene Symbol: Kdsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108342.1 | NP_001101812.1 | 9 - 10 | 954 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P576351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171927 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P576351 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P576351 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P576351 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P576351 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P100451 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93042 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83138 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10766967 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766964 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766963 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766966 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766968 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766960 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766969 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766962 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766961 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766965 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10766959 | - | - |
Rat Genome 230 2.0 Array | 1393145_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393145_at | - | - |
Affymetrix HT Rat Focus Array | 1393145_at | - | - |
Affymetrix HT Rat Focus Array | 1393145_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300008079.rn.1 | TC1300001210.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300008078.rn.1 | TC1300001210.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300008077.rn.1 | TC1300001210.rn.1 | RTA TC |
Rat Expression Set 230 | 1393145_at | - | - |
Rat Expression Set 230 | 1393145_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766959 | - | - |
Rat Genome U34 Set | rc_AA899892_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21052544 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357787 | - | - |
