Assay Details
Gene Symbol: Cdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012737.1 | NP_001012755.1 | 10 - 11 | 1868 | 135 | |
GenBank mRNA | AB121031.1 | - | 10 - 11 | 1868 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P528208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71165 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P528208 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P528208 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P528208 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P528208 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32254 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47154 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42662 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84867 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13461 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35407 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94882 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38691 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16831 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10763402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763410 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763409 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763405 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763408 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763411 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763406 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763412 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763407 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763404 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10763401 | - | - |
Rat Genome 230 2.0 Array | 1397177_at | - | - |
Rat Genome 230 2.0 Array | 1397177_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300000552.rn.1 | TC1300000109.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10763401 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372668 | - | - |
