Assay Details
Gene Symbol: Cdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012737.1 | NP_001012755.1 | 5 - 6 | 987 | 85 | |
GenBank mRNA | AB121031.1 | - | 5 - 6 | 987 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
