Assay Details
Gene Symbol: Arv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106197.1 | NP_001099667.1 | 2 - 3 | 335 | 156 | |
XM_006255806.2 | XP_006255868.1 | 2 - 3 | 293 | 156 | ||
XM_008772606.1 | XP_008770828.1 | 2 - 3 | 293 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
