Assay Details
Gene Symbol: Fa2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135583.1 | NP_001129055.1 | 2 - 3 | 435 | 90 | |
GenBank mRNA | DQ339484.1 | - | 2 - 3 | 435 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
