Assay Details
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108896.1 | NP_001102366.1 | 1 - 2 | 122 | 74 | |
GenBank mRNA | FQ228966.1 | - | 1 - 2 | 40 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108896.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
