Assay Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011927.1 | NP_001011927.1 | 6 - 7 | 895 | 85 | |
GenBank mRNA | BC088462.1 | - | 6 - 7 | 895 | 7127604 | 85 |
FQ215412.1 | - | 5 - 6 | 630 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
