Assay Details
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034146.1 | NP_001029318.1 | 9 - 10 | 1727 | 70 | |
XM_008772609.1 | XP_008770831.1 | 9 - 10 | 2060 | 70 | ||
XM_008772610.1 | XP_008770832.1 | 9 - 10 | 2055 | 70 | ||
GenBank mRNA | BC105892.1 | - | 9 - 10 | 1727 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
