Assay Details
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024257.1 | NP_001019428.1 | 3 - 4 | 300 | 89 | |
XM_006255494.2 | XP_006255556.1 | 4 - 5 | 418 | 89 | ||
XM_006255495.2 | XP_006255557.1 | 4 - 5 | 419 | 89 | ||
GenBank mRNA | BC093396.1 | - | 3 - 4 | 300 | 7388929 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
