Assay Details
Gene Symbol: Nudt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109010.1 | NP_001102480.1 | 3 - 4 | 903 | 68 | |
XM_006245588.2 | XP_006245650.1 | 3 - 4 | 908 | 68 | ||
XM_006245589.2 | XP_006245651.1 | 3 - 4 | 912 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
