Assay Details
Gene Symbol: Pja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277278.1 | NP_001264207.1 | 3 - 4 | 437 | 70 | |
NM_138896.2 | NP_620251.1 | 3 - 4 | 437 | 70 | ||
GenBank mRNA | BC074015.1 | - | 3 - 4 | 362 | 70 | |
CB577390.1 | - | 3 - 4 | 437 | 70 | ||
D32249.1 | - | 3 - 4 | 373 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
