Assay Details
Gene Symbol: Pja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138896.2 | NP_620251.1 | 4 - 5 | 1484 | 81 | |
GenBank mRNA | BC074015.1 | - | 4 - 5 | 1409 | 7105756 | 81 |
D32249.1 | - | 4 - 5 | 1420 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
