Assay Details
Gene Symbol: Efna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053903.1 | NP_446355.1 | 1 - 2 | 160 | 110 | |
XM_006245593.2 | XP_006245655.1 | 1 - 2 | 353 | 110 | ||
GenBank mRNA | U69279.1 | - | 1 - 2 | 160 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
