Assay Details
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270659.1 | NP_001257588.1 | 2 - 3 | 176 | 73 | |
NM_001276706.1 | NP_001263635.1 | 6 - 7 | 856 | 73 | ||
NM_030656.2 | NP_085914.2 | 6 - 7 | 856 | 73 | ||
GenBank mRNA | BC088133.1 | - | 6 - 7 | 688 | 73 | |
FQ228944.1 | - | 2 - 3 | 176 | 73 | ||
M35270.1 | - | 6 - 7 | 856 | 73 | ||
X06357.1 | - | 6 - 7 | 794 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
