Assay Details
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270659.1 | NP_001257588.1 | 5 - 6 | 462 | 66 | |
NM_001276706.1 | NP_001263635.1 | 9 - 10 | 1123 | 66 | ||
NM_030656.2 | NP_085914.2 | 9 - 10 | 1123 | 66 | ||
GenBank mRNA | BC088133.1 | - | 9 - 10 | 955 | 66 | |
FQ228944.1 | - | 5 - 6 | 462 | 66 | ||
M35270.1 | - | 9 - 10 | 1123 | 66 | ||
X06357.1 | - | 9 - 10 | 1061 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
