Assay Details
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276706.1 | NP_001263635.1 | 3 - 4 | 599 | 83 | |
NM_030656.2 | NP_085914.2 | 3 - 4 | 599 | 83 | ||
GenBank mRNA | BC088133.1 | - | 3 - 4 | 431 | 83 | |
M35270.1 | - | 3 - 4 | 599 | 83 | ||
X06357.1 | - | 3 - 4 | 537 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
