Assay Details
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC089788.1 | - | 118 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089788.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
