Assay Details
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008381.1 | NP_001008382.1 | 5 - 6 | 349 | 70 | |
XM_006245422.2 | XP_006245484.1 | 6 - 7 | 497 | 70 | ||
XM_006245423.2 | XP_006245485.1 | 6 - 7 | 696 | 70 | ||
XM_006245425.2 | XP_006245487.1 | 6 - 7 | 494 | 70 | ||
XM_006245427.2 | XP_006245489.1 | 6 - 7 | 541 | 70 | ||
XM_008767274.1 | XP_008765496.1 | 5 - 6 | 467 | 70 | ||
XM_008767275.1 | XP_008765497.1 | 5 - 6 | 429 | 70 | ||
GenBank mRNA | BC086355.1 | - | 5 - 6 | 349 | 7303709 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
