Assay Details
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106926.1 | NP_001100396.1 | 2 - 3 | 683 | 76 | |
XM_006245520.2 | XP_006245582.1 | 2 - 3 | 599 | 76 | ||
XM_006245521.2 | XP_006245583.1 | 2 - 3 | 457 | 76 | ||
XM_006245522.2 | XP_006245584.1 | 2 - 3 | 452 | 76 | ||
XM_008767365.1 | XP_008765587.1 | 2 - 3 | 714 | 76 | ||
GenBank mRNA | BC166559.1 | - | 1 - 2 | 356 | 7312233 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
