Assay Details
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106925.1 | NP_001100395.1 | 4 - 5 | 756 | 78 | |
XM_006245519.2 | XP_006245581.1 | 4 - 5 | 613 | 78 | ||
GenBank mRNA | BC159428.1 | - | 5 - 6 | 776 | 78 | |
FQ225176.1 | - | 4 - 5 | 619 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
