Assay Details
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057148.2 | NP_476489.1 | 2 - 3 | 74 | 151 | |
GenBank mRNA | BC081745.1 | - | 2 - 3 | 74 | 151 | |
FQ212097.1 | - | 2 - 3 | 51 | 151 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC081745.1, NM_057148.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
