Assay Details
Gene Symbol: Ppp1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009825.1 | NP_001009825.1 | 4 - 5 | 317 | 90 | |
XM_008767362.1 | XP_008765584.1 | 4 - 5 | 1263 | 90 | ||
GenBank mRNA | BC088868.1 | - | 4 - 5 | 317 | 7321115 | 90 |
FQ233576.1 | - | 331 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
