Assay Details
Gene Symbol: Ppp1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009825.1 | NP_001009825.1 | 7 - 8 | 725 | 165 | |
XM_008767362.1 | XP_008765584.1 | 7 - 8 | 1671 | 165 | ||
GenBank mRNA | BC088868.1 | - | 7 - 8 | 725 | 165 | |
FQ233576.1 | - | 739 | 165 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
