Assay Details
Gene Symbol: Hjurp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191813.1 | NP_001178742.1 | 2 - 3 | 170 | 139 | |
XM_006245393.2 | XP_006245455.1 | 2 - 3 | 321 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
