Assay Details
Gene Symbol: Iqca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767356.1 | XP_008765578.1 | 12 - 13 | 1837 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
