Assay Details
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108811.2 | NP_001102281.1 | 2 - 3 | 218 | 68 | |
XM_008767381.1 | XP_008765603.1 | 3 - 4 | 349 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
