Assay Details
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013141.2 | NP_037273.2 | 7 - 8 | 1312 | 124 | |
XM_006256166.2 | XP_006256228.1 | 7 - 8 | 2124 | 124 | ||
XM_006256167.2 | XP_006256229.1 | 6 - 7 | 2231 | 124 | ||
XM_006256168.2 | XP_006256230.1 | 6 - 7 | 3126 | 124 | ||
XM_008772797.1 | XP_008771019.1 | 6 - 7 | 2359 | 124 | ||
GenBank mRNA | AJ306400.1 | - | 5 - 6 | 1078 | 124 | |
U75918.1 | - | 7 - 8 | 1326 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
placenta development
proteoglycan metabolic process
lipid metabolic process
fatty acid beta-oxidation
vitamin A metabolic process
heart development
embryo implantation
cell population proliferation
positive regulation of cell population proliferation
axon ensheathment
phospholipid biosynthetic process
mRNA transcription
response to glucose
response to organic substance
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
negative regulation of smooth muscle cell migration
fatty acid transport
fatty acid oxidation
cell differentiation
negative regulation of cell growth
intracellular receptor signaling pathway
cell-substrate adhesion
positive regulation of insulin secretion
negative regulation of collagen biosynthetic process
response to vitamin A
wound healing
regulation of cell population proliferation
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
keratinocyte proliferation
regulation of fat cell differentiation
positive regulation of epidermis development
positive regulation of transcription, DNA-templated
decidualization
negative regulation of smooth muscle cell proliferation
negative regulation of epithelial cell proliferation
negative regulation of inflammatory response
keratinocyte migration
adipose tissue development
cellular response to hypoxia
apoptotic signaling pathway
DNA binding
DNA-binding transcription factor activity
steroid hormone receptor activity
transcription coactivator activity
nuclear receptor activity
fatty acid binding
zinc ion binding
lipid binding
sequence-specific DNA binding
protein heterodimerization activity
NF-kappaB binding
linoleic acid binding
negative regulation of transcription by RNA polymerase II
placenta development
proteoglycan metabolic process
lipid metabolic process
fatty acid beta-oxidation
vitamin A metabolic process
heart development
embryo implantation
cell population proliferation
positive regulation of cell population proliferation
axon ensheathment
phospholipid biosynthetic process
mRNA transcription
response to glucose
response to organic substance
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
negative regulation of smooth muscle cell migration
fatty acid transport
fatty acid oxidation
cell differentiation
negative regulation of cell growth
intracellular receptor signaling pathway
cell-substrate adhesion
positive regulation of insulin secretion
negative regulation of collagen biosynthetic process
response to vitamin A
wound healing
regulation of cell population proliferation
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
keratinocyte proliferation
regulation of fat cell differentiation
positive regulation of epidermis development
positive regulation of transcription, DNA-templated
decidualization
negative regulation of smooth muscle cell proliferation
negative regulation of epithelial cell proliferation
negative regulation of inflammatory response
keratinocyte migration
adipose tissue development
cellular response to hypoxia
apoptotic signaling pathway
DNA binding
DNA-binding transcription factor activity
steroid hormone receptor activity
transcription coactivator activity
nuclear receptor activity
fatty acid binding
zinc ion binding
lipid binding
sequence-specific DNA binding
protein heterodimerization activity
NF-kappaB binding
linoleic acid binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P051712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65156 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051711 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P458711 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P458711 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051711 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P051711 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P458711 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11733 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701899077 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14900 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64369 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51364 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67721 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20454 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42618 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701899077 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10828719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828717 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10828714 | - | - |
Rat Genome 230 2.0 Array | 1374914_at | - | - |
Rat Genome 230 2.0 Array | 1387684_at | - | - |
Rat Genome 230 2.0 Array | 1387684_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374914_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374914_at | - | - |
Affymetrix HT Rat Focus Array | 1387684_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387684_at | - | - |
Affymetrix HT Rat Focus Array | 1374914_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000002916.rn.1 | TC2000000312.rn.1 | RTA TC |
Rat Expression Set 230 | 1374914_PM_at | - | - |
Rat Expression Set 230 | 1387684_PM_at | - | - |
Rat Expression Set 230 | 1374914_at | - | - |
Rat Expression Set 230 | 1387684_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10828714 | - | - |
Rat Genome U34 Set | rc_AI230294_at | - | - |
Rat Genome U34 Set | U40064_g_at | - | - |
Rat Genome U34 Set | U40064_at | - | - |
Rat Genome U34 Set | U40064_g_at | - | - |
Rat Genome U34 Set | U40064_at | - | - |
Rat Genome U34 Set | rc_AI230294_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21338089 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357865 | - | - |
RatRef-12 Expression BeadChip | ILMN_53359 | - | - |
