Assay Details
Gene Symbol: Tet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256398.2 | XP_006256460.1 | 7 - 8 | 4894 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
