Assay Details
Gene Symbol: Agpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106378.1 | NP_001099848.1 | 2 - 3 | 574 | 114 | |
XM_006256243.2 | XP_006256305.1 | 3 - 4 | 369 | 114 | ||
XM_006256244.2 | XP_006256306.1 | 3 - 4 | 397 | 114 | ||
GenBank mRNA | BC158873.1 | - | 1 - 2 | 124 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
