Assay Details
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108532.1 | NP_001102002.1 | 550 | 74 | ||
XM_006256376.2 | XP_006256438.1 | 5 - 6 | 2205 | 74 | ||
XM_006256377.2 | XP_006256439.1 | 5 - 6 | 2144 | 74 | ||
XM_006256378.2 | XP_006256440.1 | 5 - 6 | 2124 | 74 | ||
XM_006256379.2 | XP_006256441.1 | 5 - 6 | 2784 | 74 | ||
XM_006256380.2 | XP_006256442.1 | 4 - 5 | 892 | 74 |
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751958.3 | XP_003752006.2 | 4 - 5 | 899 | 74 | |
XM_006256396.2 | XP_006256458.1 | 4 - 5 | 944 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
