Assay Details
Gene Symbol: Ggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053840.2 | NP_446292.2 | 1527 | 81 | ||
GenBank mRNA | BC078768.1 | - | 1527 | 81 | ||
X15443.1 | - | 1533 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
