Assay Details
Gene Symbol: Tcp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007695.1 | NP_001007696.1 | 5 - 6 | 827 | 77 | |
XM_006256200.2 | XP_006256262.1 | 5 - 6 | 686 | 77 | ||
GenBank mRNA | BC083899.1 | - | 5 - 6 | 827 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
