Assay Details
Gene Symbol: Tcp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007695.1 | NP_001007696.1 | 7 - 8 | 1181 | 61 | |
XM_006256200.2 | XP_006256262.1 | 7 - 8 | 1040 | 61 | ||
GenBank mRNA | BC083899.1 | - | 7 - 8 | 1181 | 7133508 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
