Assay Details
Gene Symbol: Bak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053812.1 | NP_446264.1 | 1 - 2 | 213 | 115 | |
GenBank mRNA | AF259504.1 | - | 1 - 2 | 213 | 115 | |
BC089784.1 | - | 1 - 2 | 174 | 7366192 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF259504.1, BC089784.1, NM_053812.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
