Assay Details
Gene Symbol: Bak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053812.1 | NP_446264.1 | 4 - 5 | 592 | 72 | |
XM_006256101.2 | XP_006256163.1 | 4 - 5 | 611 | 72 | ||
GenBank mRNA | AF259504.1 | - | 4 - 5 | 592 | 72 | |
BC089784.1 | - | 4 - 5 | 553 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
