Assay Details
Gene Symbol: Tmprss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107619.1 | NP_001101089.1 | 11 - 12 | 1402 | 80 | |
XM_006256210.2 | XP_006256272.1 | 12 - 13 | 1495 | 80 | ||
XM_006256211.2 | XP_006256273.1 | 12 - 13 | 1498 | 80 | ||
XM_008772817.1 | XP_008771039.1 | 11 - 12 | 1887 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
