Assay Details
Gene Symbol: Tmprss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107619.1 | NP_001101089.1 | 1 - 2 | 153 | 85 | |
XM_006256210.2 | XP_006256272.1 | 2 - 3 | 243 | 85 | ||
XM_006256211.2 | XP_006256273.1 | 2 - 3 | 246 | 85 | ||
XM_008772817.1 | XP_008771039.1 | 1 - 2 | 635 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
