Assay Details
Gene Symbol: Tmprss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107619.1 | NP_001101089.1 | 7 - 8 | 842 | 89 | |
XM_006256210.2 | XP_006256272.1 | 8 - 9 | 932 | 89 | ||
XM_006256211.2 | XP_006256273.1 | 8 - 9 | 935 | 89 | ||
XM_008772817.1 | XP_008771039.1 | 7 - 8 | 1324 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
