Assay Details
Gene Symbol: Mtch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100833.1 | NP_001094303.1 | 8 - 9 | 910 | 128 | |
GenBank mRNA | BC098813.1 | - | 3 - 4 | 238 | 128 | |
BC158661.1 | - | 8 - 9 | 910 | 8369713 | 128 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
