Assay Details
Gene Symbol: Rhobtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107622.1 | NP_001101092.1 | 11 - 12 | 2813 | 68 | |
XM_008772882.1 | XP_008771104.1 | 12 - 13 | 4846 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1011464 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P049311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38389 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1011464 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P049311 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P049311 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1011464 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P049311 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1011464 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P243643 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14831 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10832781 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832775 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832777 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832779 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832774 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832778 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832776 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832780 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832773 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10832772 | - | - |
Rat Genome 230 2.0 Array | 1389632_at | - | - |
Rat Genome 230 2.0 Array | 1389632_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389632_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389632_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000011686.rn.1 | TC2000001423.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000011688.rn.1 | TC2000001423.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000011687.rn.1 | TC2000001423.rn.1 | RTA TC |
Rat Expression Set 230 | 1389632_at | - | - |
Rat Expression Set 230 | 1389632_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832772 | - | - |
Rat Genome U34 Set | rc_AA925267_at | - | - |
Rat Genome U34 Set | rc_AA925267_g_at | - | - |
Rat Genome U34 Set | AA799294_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21696664 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355346 | - | - |
