Assay Details
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134417.1 | NP_599244.1 | 2 - 3 | 344 | 108 | |
XM_006256345.2 | XP_006256407.1 | 2 - 3 | 397 | 108 | ||
XM_006256346.2 | XP_006256408.1 | 2 - 3 | 759 | 108 | ||
XM_008772858.1 | XP_008771080.1 | 2 - 3 | 273 | 108 | ||
GenBank mRNA | AY014898.1 | - | 2 - 3 | 344 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
