Assay Details
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106380.1 | NP_001099850.1 | 3 - 4 | 195 | 80 | |
GenBank mRNA | BC166733.1 | - | 3 - 4 | 128 | 7936238 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
