Assay Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080891.2 | NP_543167.2 | 1 - 2 | 80 | 134 | |
XM_006256106.2 | XP_006256168.1 | 1 - 2 | 98 | 134 | ||
GenBank mRNA | BC128729.1 | - | 1 - 2 | 80 | 134 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC128729.1, NM_080891.2, XM_006256106.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
