Assay Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080891.2 | NP_543167.2 | 3 - 4 | 1126 | 98 | |
XM_006256103.2 | XP_006256165.1 | 3 - 4 | 1908 | 98 | ||
XM_006256104.2 | XP_006256166.1 | 3 - 4 | 1582 | 98 | ||
XM_006256105.2 | XP_006256167.1 | 3 - 4 | 1478 | 98 | ||
XM_006256106.2 | XP_006256168.1 | 3 - 4 | 1144 | 98 | ||
XM_006256107.2 | XP_006256169.1 | 3 - 4 | 1248 | 98 | ||
XM_006256108.2 | XP_006256170.1 | 3 - 4 | 1128 | 98 | ||
XM_006256109.1 | XP_006256171.1 | 2 - 3 | 2001 | 98 | ||
XM_006256110.2 | XP_006256172.1 | 3 - 4 | 1127 | 98 | ||
XM_008772704.1 | XP_008770926.1 | 2 - 3 | 2001 | 98 | ||
GenBank mRNA | AB064671.1 | - | 2 - 3 | 1030 | 98 | |
BC128729.1 | - | 3 - 4 | 1126 | 98 | ||
FQ223571.1 | - | 3 - 4 | 1097 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
