Assay Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080891.2 | NP_543167.2 | 6 - 7 | 1999 | 123 | |
XM_006256103.2 | XP_006256165.1 | 6 - 7 | 2781 | 123 | ||
XM_006256104.2 | XP_006256166.1 | 6 - 7 | 2455 | 123 | ||
XM_006256105.2 | XP_006256167.1 | 6 - 7 | 2351 | 123 | ||
XM_006256106.2 | XP_006256168.1 | 6 - 7 | 2017 | 123 | ||
XM_006256107.2 | XP_006256169.1 | 6 - 7 | 2121 | 123 | ||
XM_006256108.2 | XP_006256170.1 | 6 - 7 | 2001 | 123 | ||
XM_006256109.1 | XP_006256171.1 | 5 - 6 | 2874 | 123 | ||
XM_006256110.2 | XP_006256172.1 | 6 - 7 | 2000 | 123 | ||
XM_008772704.1 | XP_008770926.1 | 5 - 6 | 2874 | 123 | ||
GenBank mRNA | BC128729.1 | - | 6 - 7 | 1999 | 123 | |
FQ223571.1 | - | 6 - 7 | 1970 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
