Assay Details
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212494.2 | NP_997659.2 | 1 - 2 | 138 | 86 | |
XM_006256120.2 | XP_006256182.1 | 2 - 3 | 169 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_212494.2, XM_006256120.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
