Assay Details
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164706.1 | NP_001158178.1 | 3 - 4 | 340 | 104 | |
NM_001164707.1 | NP_001158179.1 | 2 - 3 | 393 | 104 | ||
NM_212494.2 | NP_997659.2 | 3 - 4 | 393 | 104 | ||
XM_006256120.2 | XP_006256182.1 | 4 - 5 | 424 | 104 | ||
GenBank mRNA | BP501235.1 | - | 2 - 3 | 407 | 104 | |
CB586341.1 | - | 3 - 4 | 340 | 104 | ||
DV717217.1 | - | 2 - 3 | 365 | 104 | ||
FQ220708.1 | - | 3 - 4 | 627 | 104 | ||
FQ223106.1 | - | 1 - 2 | 67 | 104 | ||
FQ229047.1 | - | 1 - 2 | 67 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
