Assay Details
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164706.1 | NP_001158178.1 | 1 - 2 | 85 | 74 | |
GenBank mRNA | CB586341.1 | - | 1 - 2 | 85 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001164706.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
