Assay Details
Gene Symbol: Gucd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134997.2 | NP_001128469.1 | 390 | 87 | ||
GenBank mRNA | BC169010.1 | - | 506 | 87 | ||
KC686830.1 | - | 506 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
